Assay Details
| Gene Symbol: LOC100390645 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752901.3 | XP_002752947.1 | 4 | 1626 | 102 | |
| XM_009004364.2 | XP_009002612.1 | 4 | 1288 | 102 | ||
More Information
Set Membership: |
|

LOC100390645
Genomic Map