Assay Details
| Gene Symbol: VSNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758017.2 | XP_002758063.1 | 4 | 2024 | 105 | |
| XM_008981111.2 | XP_008979359.1 | 4 | 1607 | 105 | ||
| XM_008981112.2 | XP_008979360.1 | 4 | 1608 | 105 | ||
More Information
Set Membership: |
|

VSNL1
Genomic Map