Assay Details
| Gene Symbol: FDFT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756766.4 | XP_002756812.1 | 8 | 2121 | 89 | |
| XM_008979026.2 | XP_008977274.1 | 7 | 1919 | 89 | ||
| XM_008979027.2 | XP_008977275.1 | 7 | 1718 | 89 | ||
| XM_008979028.2 | XP_008977276.1 | 7 | 2237 | 89 | ||
More Information
Set Membership: |
|

FDFT1
Genomic Map