Assay Details
| Gene Symbol: ARPC5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743308.4 | XP_002743354.3 | 4 | 1118 | 82 | |
| XM_017976096.1 | XP_017831585.1 | 4 | 2772 | 82 | ||
| XM_017976100.1 | XP_017831589.1 | 5 | 1256 | 82 | ||
| XM_017976104.1 | XP_017831593.1 | 4 | 1086 | 82 | ||
More Information
Set Membership: |
|

ARPC5L
Genomic Map