Assay Details
| Gene Symbol: VRK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757737.4 | XP_002757783.1 | 13 | 2022 | 86 | |
| XM_017964316.1 | XP_017819805.1 | 13 | 2034 | 86 | ||
| XM_017964317.1 | XP_017819806.1 | 13 | 1769 | 86 | ||
| XM_017964318.1 | XP_017819807.1 | 13 | 1831 | 86 | ||
More Information
Set Membership: |
|

VRK2
Genomic Map