Assay Details
| Gene Symbol: CDC42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750402.4 | XP_002750448.1 | 6 | 1313 | 80 | |
| XM_009000595.1 | XP_008998843.1 | 6 | 1286 | 80 | ||
More Information
Set Membership: |
|

CDC42
Genomic Map