Assay Details
| Gene Symbol: Il23a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172969.1 | NP_001166440.1 | 2 - 3 | 257 | 66 | |
| GenBank mRNA | AB058509.1 | - | 2 - 3 | 257 | 66 | |
More Information
Set Membership: |
|

Il23a
Genomic Map