Assay Details
| Gene Symbol: Gnrh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172956.1 | NP_001166427.1 | 2 - 3 | 255 | 99 | |
| XM_005007999.2 | XP_005008056.1 | 1 - 2 | 158 | 99 | ||
| GenBank mRNA | AF033346.1 | - | 2 - 3 | 255 | 99 | |
More Information
Set Membership: |
|

Gnrh1
Genomic Map