Assay Details
| Gene Symbol: Ramp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173009.1 | NP_001166480.1 | 1 - 2 | 146 | 53 | |
| GenBank mRNA | AF484219.1 | - | 1 - 2 | 146 | 53 | |
More Information
Set Membership: |
|

Ramp1
Genomic Map