Assay Details
| Gene Symbol: Leap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003464467.4 | XP_003464515.2 | 2 - 3 | 84 | ||
| GenBank mRNA | AJ409054.1 | - | 2 - 3 | 180 | 84 | |
More Information
Set Membership: |
|

Leap2
Genomic Map