Assay Details
| Gene Symbol: Hsd11b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172857.1 | NP_001166328.1 | 5 - 6 | 714 | 82 | |
| XM_013157260.2 | XP_013012714.1 | 6 - 7 | 747 | 82 | ||
| GenBank mRNA | AF188005.1 | - | 5 - 6 | 714 | 82 | |
| AY535424.1 | - | 5 - 6 | 667 | 82 | ||
More Information
Set Membership: |
|

Hsd11b1
Genomic Map