Assay Details
| Gene Symbol: Ptger4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173056.1 | NP_001166527.1 | 1 - 2 | 923 | 66 | |
| GenBank mRNA | DQ489549.1 | - | 1 - 2 | 923 | 66 | |
More Information
Set Membership: |
|

Ptger4
Genomic Map