Assay Details
| Gene Symbol: Iapp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172965.2 | NP_001166436.1 | 2 - 3 | 200 | 67 | |
| GenBank mRNA | M25387.1 | - | 2 - 3 | 201 | 67 | |
More Information
Set Membership: |
|

Iapp
Genomic Map