Assay Details
| Gene Symbol: F9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003464707.2 | XP_003464755.1 | 6 - 7 | 788 | 67 | |
| GenBank mRNA | M26237.1 | - | 1 - 2 | 219 | 67 | |
More Information
Set Membership: |
|

F9
Genomic Map