Assay Details
| Gene Symbol: Crisp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172934.1 | NP_001166405.1 | 4 - 5 | 250 | 64 | |
| GenBank mRNA | U35712.1 | - | 4 - 5 | 250 | 64 | |
More Information
Set Membership: |
|

Crisp2
Genomic Map