Assay Details
| Gene Symbol: Csf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003464529.2 | XP_003464577.1 | 2 - 3 | 511 | 79 | |
| GenBank mRNA | U46779.1 | - | 2 - 3 | 201 | 79 | |
More Information
Set Membership: |
|

Csf2
Genomic Map