Assay Details
| Gene Symbol: Gapdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172951.1 | NP_001166422.1 | 8 - 9 | 731 | 65 | |
| GenBank mRNA | AB060340.1 | - | 1 | 665 | 65 | |
| DQ403052.1 | - | 1 | 597 | 65 | ||
| EU862201.1 | - | 8 - 9 | 731 | 65 | ||
| U51572.1 | - | 1 | 178 | 65 | ||
| U76741.1 | - | 1 - 2 | 71 | 65 | ||
More Information
Set Membership: |
|

Gapdh
Genomic Map