Assay Details
| Gene Symbol: Ca3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173075.1 | NP_001166546.1 | 5 - 6 | 547 | 70 | |
| GenBank mRNA | EU327786.1 | - | 5 - 6 | 547 | 70 | |
More Information
Set Membership: |
|

Ca3
Genomic Map