Assay Details
| Gene Symbol: Gnrhr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172957.2 | NP_001166428.1 | 1 - 2 | 572 | 70 | |
| GenBank mRNA | AF426176.3 | - | 1 - 2 | 573 | 70 | |
More Information
Set Membership: |
|

Gnrhr
Genomic Map