Assay Details
| Gene Symbol: Gpr34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172749.1 | NP_001166220.1 | 1 | 1046 | 73 | |
| XM_013151875.2 | XP_013007329.1 | 4 | 1431 | 73 | ||
| XM_013151876.2 | XP_013007330.1 | 4 | 1428 | 73 | ||
| GenBank mRNA | AB159442.1 | - | 1 | 1046 | 73 | |
More Information
Set Membership: |
|

Gpr34
Genomic Map