Assay Details
| Gene Symbol: Crybb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173071.1 | NP_001166542.1 | 3 - 4 | 227 | 75 | |
| GenBank mRNA | EB370475.1 | - | 3 - 4 | 217 | 75 | |
| EF457998.1 | - | 3 - 4 | 227 | 75 | ||
More Information
Set Membership: |
|

Crybb2
Genomic Map