Assay Details
| Gene Symbol: Gstm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172834.1 | NP_001166305.1 | 6 - 7 | 528 | 63 | |
| GenBank mRNA | AB000488.1 | - | 6 - 7 | 528 | 63 | |
More Information
Set Membership: |
|

Gstm2
Genomic Map