Assay Details
| Gene Symbol: Hprt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003462671.4 | XP_003462719.2 | 2 - 3 | 331 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hprt1| Gene Symbol: Hprt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003462671.4 | XP_003462719.2 | 2 - 3 | 331 | 73 | |
Set Membership: |
|