Assay Details
| Gene Symbol: CG9426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135772.2 | NP_609616.1 | 2 - 3 | 1079 | 63 | |
| GenBank mRNA | AY118728.1 | - | 2 - 3 | 650 | 63 | |
More Information
Set Membership: |
|

CG9426
Genomic Map