Assay Details
| Gene Symbol: CG9426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135772.2 | NP_609616.1 | 4 - 5 | 2182 | 68 | |
| GenBank mRNA | AY118728.1 | - | 4 - 5 | 1753 | 68 | |
More Information
Set Membership: |
|

CG9426
Genomic Map