Assay Details
| Gene Symbol: Neu2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141059.3 | NP_649316.1 | 2 | 1248 | 61 | |
| GenBank mRNA | BT028817.2 | - | 1 | 1108 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Neu2| Gene Symbol: Neu2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141059.3 | NP_649316.1 | 2 | 1248 | 61 | |
| GenBank mRNA | BT028817.2 | - | 1 | 1108 | 61 | |
Set Membership: |
|