Assay Details
| Gene Symbol: Est-Q | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141064.3 | NP_649321.1 | 2 - 3 | 447 | 72 | |
| GenBank mRNA | AY051599.1 | - | 2 - 3 | 412 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Est-Q| Gene Symbol: Est-Q | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141064.3 | NP_649321.1 | 2 - 3 | 447 | 72 | |
| GenBank mRNA | AY051599.1 | - | 2 - 3 | 412 | 72 | |
Set Membership: |
|