Assay Details
| Gene Symbol: MED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202224.2 | NP_001189153.1 | 2 - 3 | 1467 | 61 | |
| NM_001275254.1 | NP_001262183.1 | 2 - 3 | 1467 | 61 | ||
| NM_141084.3 | NP_649341.1 | 2 - 3 | 1467 | 61 | ||
| GenBank mRNA | AF289996.1 | - | 2 - 3 | 1455 | 61 | |
| BT044447.1 | - | 2 - 3 | 1419 | 61 | ||
| BT133280.1 | - | 2 - 3 | 1467 | 61 | ||
More Information
Set Membership: |
|

MED1
Genomic Map