Assay Details
| Gene Symbol: wal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057627.4 | NP_476975.1 | 3 - 4 | 255 | 64 | |
| NM_165847.2 | NP_725073.1 | 3 - 4 | 255 | 64 | ||
| GenBank mRNA | AF132155.1 | - | 3 - 4 | 238 | 64 | |
More Information
Set Membership: |
|

wal
Genomic Map