Assay Details
| Gene Symbol: Spt-I | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136998.2 | NP_610842.1 | 2 - 3 | 607 | 68 | |
| NM_165967.4 | NP_725255.1 | 3 - 4 | 679 | 68 | ||
| NM_165968.4 | NP_725256.1 | 3 - 4 | 649 | 68 | ||
| GenBank mRNA | BT010234.1 | - | 3 - 4 | 647 | 68 | |
More Information
Set Membership: |
|

Spt-I
Genomic Map