Assay Details
| Gene Symbol: Dgk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103749.2 | NP_001097219.1 | 19 - 20 | 3942 | 74 | |
| NM_001103750.3 | NP_001097220.1 | 18 - 19 | 3633 | 74 | ||
| NM_001273863.1 | NP_001260792.1 | 18 - 19 | 3609 | 74 | ||
| NM_001316408.1 | NP_001303337.1 | 18 - 19 | 3609 | 74 | ||
| NM_078930.4 | NP_523654.2 | 8 - 9 | 2547 | 74 | ||
| NM_165568.3 | NP_724619.2 | 20 - 21 | 3999 | 74 | ||
| NM_206053.2 | NP_995775.2 | 7 - 8 | 2415 | 74 | ||
| GenBank mRNA | BT003323.1 | - | 8 - 9 | 2255 | 74 | |
More Information
Set Membership: |
|

Dgk
Genomic Map