Assay Details
| Gene Symbol: PCNA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057557.4 | NP_476905.1 | 1 - 2 | 267 | 76 | |
| NM_206182.2 | NP_995904.1 | 1 - 2 | 194 | 76 | ||
| GenBank mRNA | AY122197.1 | - | 1 - 2 | 222 | 76 | |
More Information
Set Membership: |
|

PCNA
Genomic Map