Assay Details
| Gene Symbol: RpL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057283.5 | NP_476631.1 | 2 - 3 | 189 | 82 | |
| NM_206219.2 | NP_995941.1 | 2 - 3 | 151 | 82 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RpL19| Gene Symbol: RpL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057283.5 | NP_476631.1 | 2 - 3 | 189 | 82 | |
| NM_206219.2 | NP_995941.1 | 2 - 3 | 151 | 82 | ||
Set Membership: |
|