Assay Details
| Gene Symbol: nSyb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259618.2 | NP_001246547.1 | 5 - 6 | 852 | 69 | |
| NM_001274340.1 | NP_001261269.1 | 4 - 5 | 791 | 69 | ||
| NM_001274341.1 | NP_001261270.1 | 4 - 5 | 803 | 69 | ||
| NM_057710.6 | NP_477058.1 | 4 - 5 | 779 | 69 | ||
| NM_167904.3 | NP_728644.1 | 4 - 5 | 782 | 69 | ||
| NM_167905.4 | NP_728645.1 | 5 - 6 | 855 | 69 | ||
| NM_167906.3 | NP_728646.2 | 5 - 6 | 852 | 69 | ||
| NM_206234.4 | NP_995956.1 | 5 - 6 | 852 | 69 | ||
| GenBank mRNA | AY069045.1 | - | 3 - 4 | 760 | 69 | |
| BT099571.1 | - | 4 - 5 | 830 | 69 | ||
More Information
Set Membership: |
|

nSyb
Genomic Map