Assay Details
| Gene Symbol: enc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104026.2 | NP_001097496.1 | 6 - 7 | 2188 | 103 | |
| NM_001104027.2 | NP_001097497.1 | 6 - 7 | 2131 | 103 | ||
| NM_001274447.1 | NP_001261376.1 | 6 - 7 | 2146 | 103 | ||
| NM_001274448.1 | NP_001261377.1 | 5 - 6 | 1688 | 103 | ||
| NM_001274449.1 | NP_001261378.1 | 6 - 7 | 2131 | 103 | ||
| NM_080026.3 | NP_524765.2 | 6 - 7 | 2131 | 103 | ||
| NM_206270.2 | NP_995992.1 | 6 - 7 | 2146 | 103 | ||
| GenBank mRNA | BT050449.1 | - | 6 - 7 | 2129 | 103 | |
More Information
Set Membership: |
|

enc
Genomic Map