Assay Details
| Gene Symbol: NAAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131991.3 | NP_572219.1 | 3 - 4 | 919 | 64 | |
| GenBank mRNA | BT099637.1 | - | 3 - 4 | 915 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NAAT1| Gene Symbol: NAAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131991.3 | NP_572219.1 | 3 - 4 | 919 | 64 | |
| GenBank mRNA | BT099637.1 | - | 3 - 4 | 915 | 64 | |
Set Membership: |
|