Assay Details
| Gene Symbol: NAAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131991.3 | NP_572219.1 | 6 - 7 | 1938 | 126 | |
| GenBank mRNA | BT099637.1 | - | 6 - 7 | 1934 | 126 | |
More Information
Set Membership: |
|

NAAT1
Genomic Map