Assay Details
| Gene Symbol: NAAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131991.3 | NP_572219.1 | 7 - 8 | 2030 | 65 | |
| GenBank mRNA | BT099637.1 | - | 7 - 8 | 2026 | 65 | |
More Information
Set Membership: |
|

NAAT1
Genomic Map