Assay Details
| Gene Symbol: mEFTs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136003.5 | NP_609847.1 | 2 - 3 | 858 | 57 | |
| GenBank mRNA | AY051938.1 | - | 2 - 3 | 825 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mEFTs| Gene Symbol: mEFTs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136003.5 | NP_609847.1 | 2 - 3 | 858 | 57 | |
| GenBank mRNA | AY051938.1 | - | 2 - 3 | 825 | 57 | |
Set Membership: |
|