Assay Details
| Gene Symbol: Atg10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103745.3 | NP_001097215.1 | 4 | 411 | 68 | |
| NM_001103746.3 | NP_001097216.2 | 4 | 411 | 68 | ||
| GenBank mRNA | BT022398.1 | - | 4 | 383 | 68 | |
| BT088901.1 | - | 4 | 383 | 68 | ||
More Information
Set Membership: |
|

Atg10
Genomic Map