Assay Details
| Gene Symbol: Rcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274033.2 | NP_001260962.1 | 1 - 2 | 278 | 91 | |
| NM_001274035.2 | NP_001260964.1 | 1 - 2 | 278 | 91 | ||
| NM_166031.3 | NP_725357.2 | 1 - 2 | 278 | 91 | ||
| GenBank mRNA | BT100232.1 | - | 1 - 2 | 244 | 91 | |
More Information
Set Membership: |
|

Rcd1
Genomic Map