Assay Details
| Gene Symbol: Rcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137083.4 | NP_610927.3 | 5 - 6 | 2167 | 67 | |
| NM_166031.3 | NP_725357.2 | 5 - 6 | 1800 | 67 | ||
| NM_166032.3 | NP_725358.1 | 5 - 6 | 1637 | 67 | ||
| GenBank mRNA | AY069502.1 | - | 5 - 6 | 2142 | 67 | |
| BT100232.1 | - | 5 - 6 | 1766 | 67 | ||
More Information
Set Membership: |
|

Rcd1
Genomic Map