Assay Details
| Gene Symbol: scb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079026.3 | NP_523750.2 | 2 - 3 | 1694 | 67 | |
| NM_166083.2 | NP_725445.1 | 2 - 3 | 1355 | 67 | ||
| GenBank mRNA | BT015245.1 | - | 2 - 3 | 1677 | 67 | |
| BT021944.1 | - | 2 - 3 | 1327 | 67 | ||
| U76605.1 | - | 2 - 3 | 1682 | 67 | ||
More Information
Set Membership: |
|

scb
Genomic Map