Assay Details
| Gene Symbol: Strn-Mlck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202023.2 | NP_001188952.1 | 5 - 6 | 1627 | 58 | |
| NM_001202024.2 | NP_001188953.2 | 5 - 6 | 1662 | 58 | ||
| NM_001202026.2 | NP_001188955.2 | 23 - 24 | 23638 | 58 | ||
| NM_001274089.1 | NP_001261018.1 | 23 - 24 | 23638 | 58 | ||
| NM_001274090.1 | NP_001261019.1 | 23 - 24 | 24006 | 58 | ||
| NM_079030.4 | NP_523754.2 | 5 - 6 | 1616 | 58 | ||
| NM_166128.3 | NP_725509.2 | 4 - 5 | 1677 | 58 | ||
| NM_166129.3 | NP_725510.2 | 23 - 24 | 24006 | 58 | ||
More Information
Set Membership: |
|

Strn-Mlck
Genomic Map