Assay Details
| Gene Symbol: Dg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079032.4 | NP_523756.1 | 6 - 7 | 1317 | 78 | |
| NM_166138.5 | NP_725523.3 | 6 - 7 | 1317 | 78 | ||
| NM_176183.3 | NP_788363.1 | 6 - 7 | 1317 | 78 | ||
| NM_176184.4 | NP_788364.2 | 6 - 7 | 1317 | 78 | ||
| GenBank mRNA | AF461100.1 | - | 5 - 6 | 669 | 78 | |
| AF461101.1 | - | 5 - 6 | 669 | 78 | ||
| AY069076.1 | - | 6 - 7 | 1290 | 78 | ||
More Information
Set Membership: |
|

Dg
Genomic Map