Assay Details
| Gene Symbol: Dg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079032.4 | NP_523756.1 | 9 - 10 | 1676 | 73 | |
| NM_166138.5 | NP_725523.3 | 8 - 9 | 1427 | 73 | ||
| NM_176183.3 | NP_788363.1 | 10 - 11 | 2471 | 73 | ||
| NM_176184.4 | NP_788364.2 | 9 - 10 | 2222 | 73 | ||
| GenBank mRNA | AF461100.1 | - | 8 - 9 | 1574 | 73 | |
| AF461101.1 | - | 8 - 9 | 1028 | 73 | ||
| AY069076.1 | - | 9 - 10 | 1649 | 73 | ||
More Information
Set Membership: |
|

Dg
Genomic Map