Assay Details
| Gene Symbol: csul | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057836.4 | NP_477184.1 | 2 - 3 | 931 | 66 | |
| NM_166158.2 | NP_725552.1 | 2 - 3 | 916 | 66 | ||
| GenBank mRNA | AY122153.1 | - | 2 - 3 | 899 | 66 | |
More Information
Set Membership: |
|

csul
Genomic Map