Assay Details
| Gene Symbol: MsrA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274949.1 | NP_001261878.1 | 3 - 4 | 644 | 68 | |
| NM_001274950.1 | NP_001261879.1 | 3 - 4 | 568 | 68 | ||
| NM_079361.3 | NP_524085.2 | 3 - 4 | 568 | 68 | ||
| NM_168622.2 | NP_730047.1 | 3 - 4 | 556 | 68 | ||
| NM_168623.2 | NP_730048.1 | 2 - 3 | 678 | 68 | ||
| NM_206371.3 | NP_996093.2 | 3 - 4 | 520 | 68 | ||
| GenBank mRNA | AF541958.1 | - | 3 - 4 | 501 | 68 | |
| BT044581.1 | - | 3 - 4 | 567 | 68 | ||
More Information
Set Membership: |
|

MsrA
Genomic Map