Assay Details
| Gene Symbol: svr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297814.1 | NP_001284743.1 | 1 - 2 | 651 | 57 | |
| NM_001297815.1 | NP_001284744.1 | 1 - 2 | 651 | 57 | ||
| NM_080293.4 | NP_525032.2 | 2 - 3 | 930 | 57 | ||
| NM_176679.3 | NP_788852.2 | 1 - 2 | 651 | 57 | ||
| NM_206596.2 | NP_996319.1 | 2 - 3 | 930 | 57 | ||
| NM_206597.2 | NP_996320.1 | 2 - 3 | 930 | 57 | ||
| GenBank mRNA | AF545817.1 | - | 1 - 2 | 708 | 57 | |
| AF545818.1 | - | 1 - 2 | 708 | 57 | ||
| AF545820.1 | - | 1 - 2 | 708 | 57 | ||
| BT099720.1 | - | 1 - 2 | 621 | 57 | ||
| BT100053.1 | - | 1 - 2 | 622 | 57 | ||
More Information
Set Membership: |
|

svr
Genomic Map