Assay Details
| Gene Symbol: svr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144669.2 | NP_001138141.1 | 2 - 3 | 1255 | 57 | |
| NM_001297813.1 | NP_001284742.1 | 3 - 4 | 1130 | 57 | ||
| NM_001297814.1 | NP_001284743.1 | 2 - 3 | 889 | 57 | ||
| NM_001297815.1 | NP_001284744.1 | 2 - 3 | 889 | 57 | ||
| NM_080293.4 | NP_525032.2 | 3 - 4 | 1168 | 57 | ||
| NM_166846.5 | NP_726675.3 | 2 - 3 | 1182 | 57 | ||
| NM_176679.3 | NP_788852.2 | 2 - 3 | 889 | 57 | ||
| NM_206596.2 | NP_996319.1 | 3 - 4 | 1168 | 57 | ||
| NM_206597.2 | NP_996320.1 | 3 - 4 | 1168 | 57 | ||
| NM_206598.3 | NP_996321.1 | 2 - 3 | 1182 | 57 | ||
| NM_206599.2 | NP_996322.1 | 2 - 3 | 1182 | 57 | ||
| GenBank mRNA | AF545816.1 | - | 2 - 3 | 826 | 57 | |
| AF545817.1 | - | 2 - 3 | 946 | 57 | ||
| AF545818.1 | - | 2 - 3 | 946 | 57 | ||
| AF545819.1 | - | 2 - 3 | 826 | 57 | ||
| AF545820.1 | - | 2 - 3 | 946 | 57 | ||
| BT015253.1 | - | 2 - 3 | 825 | 57 | ||
| BT099720.1 | - | 2 - 3 | 859 | 57 | ||
| BT100053.1 | - | 2 - 3 | 860 | 57 | ||
| BT100310.1 | - | 2 - 3 | 368 | 57 | ||
More Information
Set Membership: |
|

svr
Genomic Map